KEY428741972 Key Organics / BIONET

1-{[3-(3-phenyl-1,2,4-oxadiazol-5-yl)pyridin-2-yl]amino}pyrrolidine-2,5-dione

  • MDL number MFCD14703590
  • Cas Number : 1232799-90-4
  • Molecular Weight : 335.323

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440260

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3354_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart