KEY428741973 Key Organics / BIONET

1-(4-ethylphenyl)-4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD06191747
  • Cas Number : 727374-25-6
  • Molecular Weight : 252.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440261

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3355_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart