KEY428741974 Key Organics / BIONET

(2E)-3-(2-hydroxyphenyl)-1-(4-methylphenyl)prop-2-en-1-one

  • MDL number MFCD00225999
  • Cas Number : 7645-95-6
  • Molecular Weight : 238.286

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440262

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3358_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart