KEY428741976 Key Organics / BIONET

ethyl 4-amino-2-sulfanylidene-3-[3-(trifluoromethyl)phenyl]-2,3-dihydro-1,3-thiazole-5-carboxylate

  • MDL number MFCD02089347
  • Cas Number : 312922-28-4
  • Molecular Weight : 348.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440264

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3360_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart