KEY428741978 Key Organics / BIONET

2-(1H-1,3-benzodiazol-2-yl)-3-(4-chlorophenyl)propanethioamide

  • MDL number MFCD18203424
  • Cas Number : 1260920-39-5
  • Molecular Weight : 315.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440266

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3362_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart