KEY428741980 Key Organics / BIONET

2-(1H-1,3-benzodiazol-2-yl)-3-(4-methoxyphenyl)propanethioamide

  • MDL number MFCD18203426
  • Cas Number : 1260918-96-4
  • Molecular Weight : 311.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440268

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3364_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart