KEY428741982 Key Organics / BIONET

2-(2-methylphenyl)-4H,5H-pyrazolo[1,5-a]pyrazin-4-one

  • MDL number MFCD24850924
  • Cas Number : 1338664-59-7
  • Molecular Weight : 225.251

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440270

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3366_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart