KEY428741983 Key Organics / BIONET

2,2-dimethyl-4-phenyl-2,5-dihydro-1H-imidazole-5-thione

  • MDL number MFCD02311186
  • Cas Number : 4602-37-3
  • Molecular Weight : 204.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440271

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3368_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart