KEY428741984 Key Organics / BIONET

ethyl 2-amino-4-methyl-5-(morpholine-4-carbonyl)thiophene-3-carboxylate

  • MDL number MFCD00718001
  • Cas Number : 314046-67-8
  • Molecular Weight : 298.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440272

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3369_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart