KEY428741985 Key Organics / BIONET

3-(4-chlorophenyl)-1,4-diazaspiro[4.5]dec-3-en-2-one

  • MDL number MFCD14735655
  • Cas Number : 923975-81-9
  • Molecular Weight : 262.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440273

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3371_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart