KEY428741994 Key Organics / BIONET

methyl 3-(3-aminophenyl)-2-acetamidopropanoate

  • MDL number MFCD28122602
  • Cas Number : 219826-11-6
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199 - 200
Purity 0.97
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440282

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/FK-0719_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart