KEY428742000 Key Organics / BIONET

methyl {[3-(3-methoxyphenyl)-1,2-oxazol-5-yl]carbamoyl}formate

  • MDL number MFCD28122606
  • Cas Number : 1622069-62-8
  • Molecular Weight : 276.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150 - 152
Purity 0.97
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440288

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/HK-0216_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart