KEY428742045 Key Organics / BIONET

(Z)-2-[(1-bromonaphthalen-2-yl)oxy]-N'-hydroxyethanimidamide

  • MDL number MFCD00245486
  • Cas Number : 884504-67-0
  • Molecular Weight : 295.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/105803

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/MS-7101_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart