KEY428742060 Key Organics / BIONET

2,6,6-trimethyl-4,5,6,7-tetrahydro-1H-indol-4-one

  • MDL number MFCD00129627
  • Cas Number : 69595-03-5
  • Molecular Weight : 177.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Dark Orange Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/106278

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/MS-7674_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart