KEY428742065 Key Organics / BIONET

1-(4-chlorophenyl)-3-[(pyridin-3-yl)methyl]urea

  • MDL number MFCD00023607
  • Cas Number : 53101-97-6
  • Molecular Weight : 261.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/106570

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/MS-8026_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart