KEY428742070 Key Organics / BIONET

(2E)-3-(3-cyanophenyl)prop-2-enoic acid

  • MDL number MFCD00182407
  • Cas Number : 16642-93-6
  • Molecular Weight : 173.171

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/430195

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/MS-8957_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart