KEY428742079 Key Organics / BIONET

ethyl 2-(1-benzyl-3-oxopiperazin-2-yl)acetate

  • MDL number MFCD13250153
  • Cas Number : 61155-09-7
  • Molecular Weight : 276.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440290

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-3070_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart