KEY428742083 Key Organics / BIONET

3,5-dimethyl-N-(propan-2-yl)aniline

  • MDL number MFCD11145897
  • Cas Number : 52827-66-4
  • Molecular Weight : 163.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Colourless to Dark Brown Liquid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440293

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4847_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart