KEY428742086 Key Organics / BIONET

1,3,5-trimethyl-1,2-dihydropyridin-2-one

  • MDL number MFCD28097869
  • Cas Number : 98042-58-1
  • Molecular Weight : 137.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440296

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4850_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart