KEY429323981 Key Organics / BIONET

2-{4-[(4-methoxyphenyl)methyl]phenyl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD28127159
  • Cas Number : 2052955-21-0
  • Molecular Weight : 324.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-(4-methoxybenzyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Inchi Code 1S/C20H25BO3/c1-19(2)20(3,4)24-21(23-19)17-10-6-15(7-11-17)14-16-8-12-18(22-5)13-9-16/h6-13H,14H2,1-5H3
Inchi Key QFRIXBSWUHSRMS-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440865

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3087_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart