KEY429323983 Key Organics / BIONET

potassium [4-(3-carboxypropanamido)phenyl]trifluoroboranuide

  • MDL number MFCD28127161
  • Molecular Weight : 299.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440867

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-3089_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart