KEY429323985 Key Organics / BIONET

4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]thiomorpholine

  • MDL number MFCD18761784
  • Cas Number : 1352413-75-2
  • Molecular Weight : 306.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440858

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3080_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart