KEY429323988 Key Organics / BIONET

6-bromo-2H-chromen-2-one

  • MDL number MFCD01327473
  • Cas Number : 19063-55-9
  • Molecular Weight : 225.041

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440857

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-3079_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart