KEY429323989 Key Organics / BIONET

4-(2,2,4-trimethyl-3,4-dihydro-2H-1-benzopyran-4-yl)phenol

  • MDL number MFCD00033694
  • Cas Number : 472-41-3
  • Molecular Weight : 268.356

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440861

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-3083_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart