KEY429323995 Key Organics / BIONET

3-amino-1,3-oxazolidin-2-one hydrochloride

  • MDL number MFCD04038842
  • Cas Number : 5567-67-9
  • Molecular Weight : 138.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126 - 129
Purity 0.95
Storage temp. Store under N2, Hygroscopic
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440873

Safety Information

signalword Warning
pcodes P232,P260,P270,P280,P402 + P404,P422
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-7039_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart