KEY429323997 Key Organics / BIONET

2-chloro-N-(1H-1,2,4-triazol-5-yl)acetamide

  • MDL number MFCD17780147
  • Cas Number : 52938-49-5
  • Molecular Weight : 160.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440875

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3321_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart