KEY429323998 Key Organics / BIONET

1-[2-(3,4-dimethoxyphenyl)pyrazolo[1,5-a]pyrazin-4-yl]piperidine-4-carboxylic acid

  • MDL number MFCD18201617
  • Cas Number : 1111058-13-9
  • Molecular Weight : 382.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440876

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3322_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart