KEY429324000 Key Organics / BIONET

1,3-bis(1-benzyl-5-oxopyrrolidin-3-yl)urea

  • MDL number MFCD28041400
  • Cas Number : 1370593-69-3
  • Molecular Weight : 406.486

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440878

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3340_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart