KEY429324001 Key Organics / BIONET

2-phenylcyclopropane-1-carbohydrazide

  • MDL number MFCD00462672
  • Cas Number : 14814-55-2
  • Molecular Weight : 176.219

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 88
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440879

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3370_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart