KEY429324004 Key Organics / BIONET

3,5-dichloro-1-[(4-methoxyphenyl)methyl]-1,2-dihydropyrazin-2-one

  • MDL number MFCD24683345
  • Cas Number : 393860-82-7
  • Molecular Weight : 285.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440882

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-0600_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart