KEY429324015 Key Organics / BIONET

7-methyl-1H,2H,3H,4H-thieno[3,2-d]pyrimidine-2,4-dione

  • MDL number MFCD20617523
  • Cas Number : 35265-81-7
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440893

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GS-0611_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart