KEY429324024 Key Organics / BIONET

2,4-dichloro-7,7-dimethyl-5,6,7,8-tetrahydroquinolin-5-one

  • MDL number MFCD28127162
  • Cas Number : 1630096-70-6
  • Molecular Weight : 244.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440902

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GS-0620_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart