KEY429324025 Key Organics / BIONET

6-chloro-8-methyl-1,2,3,4-tetrahydroquinazoline-2,4-dione

  • MDL number MFCD24614691
  • Cas Number : 1080622-94-1
  • Molecular Weight : 210.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440903

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GS-0621_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart