KEY429324028 Key Organics / BIONET

methyl 4-amino-5,5-dimethyl-2,5-dihydrofuran-3-carboxylate

  • MDL number MFCD19703786
  • Cas Number : 1260088-70-7
  • Molecular Weight : 171.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440906

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GS-0624_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart