KEY429324035 Key Organics / BIONET

1-benzyl-4-bromopiperidine

  • MDL number MFCD16496496
  • Cas Number : 301665-60-1
  • Molecular Weight : 254.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440913

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-0631_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart