KEY429324042 Key Organics / BIONET

7,7-dimethyl-1,2,5,6,7,8-hexahydroquinoline-2,5-dione

  • MDL number MFCD09837269
  • Cas Number : 55119-00-1
  • Molecular Weight : 191.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440920

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GS-0638_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart