KEY429324047 Key Organics / BIONET

2-amino-N-[(pyridin-2-yl)methyl]ethane-1-sulfonamide hydrochloride

  • MDL number MFCD28127029
  • Cas Number : 1630096-72-8
  • Molecular Weight : 251.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440925

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GS-0643_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart