KEY429324048 Key Organics / BIONET

ethyl 4-amino-5,5-dimethyl-2-oxo-2,5-dihydrofuran-3-carboxylate

  • MDL number MFCD18447452
  • Cas Number : 139003-76-2
  • Molecular Weight : 199.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440926

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GS-0644_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart