KEY429324049 Key Organics / BIONET

1-{3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy]propyl}piperidine

  • MDL number MFCD09475821
  • Cas Number : 401895-68-9
  • Molecular Weight : 345.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440927

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GS-0645_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart