KEY429324057 Key Organics / BIONET

2-bromo-6-(chloromethyl)pyridine

  • MDL number MFCD10697585
  • Cas Number : 727356-19-6
  • Molecular Weight : 206.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 46 - 49
Iupac Name 2-bromo-6-(chloromethyl)pyridine
Inchi Code 1S/C6H5BrClN/c7-6-3-1-2-5(4-8)9-6/h1-3H,4H2
Inchi Key BYQJMBHZLIDFBY-UHFFFAOYSA-N

Description

Melting Point 46 - 49
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440936

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/HK-0028_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart