KEY429324062 Key Organics / BIONET

methyl (2S)-2-acetamido-3-[4-(4-methylpiperazin-1-yl)phenyl]propanoate

  • MDL number MFCD28127032
  • Cas Number : 1630114-56-5
  • Molecular Weight : 319.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108 - 109
Purity 97
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440941

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/HK-0703_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart