KEY429324064 Key Organics / BIONET

4-[(E)-[(2H-1,3-benzodioxol-5-yl)methylidene]amino]phenol

  • MDL number MFCD00441530
  • Cas Number : 51209-73-5
  • Molecular Weight : 241.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440943

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-2977_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart