KEY429324065 Key Organics / BIONET

(2E)-3-(3,4-dichlorophenyl)-1-(4-hydroxyphenyl)prop-2-en-1-one

  • MDL number MFCD01935363
  • Cas Number : 204974-31-2
  • Molecular Weight : 293.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440944

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-2979_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart