KEY429324066 Key Organics / BIONET

2-(2,2-dimethylpropanamido)benzoic acid

  • MDL number MFCD01006384
  • Cas Number : 101724-84-9
  • Molecular Weight : 221.256

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440945

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/JS-2981_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart