KEY429324069 Key Organics / BIONET

(2Z)-1-(4-hydroxyphenyl)-3-(naphthalen-1-yl)prop-2-en-1-one

  • MDL number MFCD28127034
  • Cas Number : 385810-16-2
  • Molecular Weight : 274.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440948

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-2995_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart