KEY432310512 Key Organics / BIONET

2-{4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperazin-1-yl}ethan-1-amine

  • MDL number MFCD28139663
  • Cas Number : 1489230-58-1
  • Molecular Weight : 332.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440863

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-3085_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart