KEY432310527 Key Organics / BIONET

(E)-2-(3,4-difluorophenyl)ethene-1-sulfonyl chloride

  • MDL number MFCD04973465
  • Cas Number : 1158108-22-5
  • Molecular Weight : 238.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441452

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-7066_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart