KEY432310528 Key Organics / BIONET

3-[(4-phenylpiperazin-1-yl)sulfonyl]-1,2-dihydropyridin-2-one

  • MDL number MFCD19105022
  • Cas Number : 1325306-66-8
  • Molecular Weight : 319.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441453

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3372_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart