KEY432310530 Key Organics / BIONET

3-[(4-methylpiperidin-1-yl)sulfonyl]-1,2-dihydropyridin-2-one

  • MDL number MFCD19104878
  • Cas Number : 1325306-17-9
  • Molecular Weight : 256.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441455

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3374_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart