KEY432310534 Key Organics / BIONET

3-ethyl-N-(3-fluorophenyl)-[1,2,4]triazolo[4,3-a]pyridine-8-sulfonamide

  • MDL number MFCD18804107
  • Cas Number : 1291486-53-7
  • Molecular Weight : 320.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441459

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3378_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart